Assay Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278610.1 | NP_001265539.1 | 8 - 9 | 1699 | 66 | |
| NM_002466.3 | NP_002457.1 | 9 - 10 | 1771 | 66 | ||
| GenBank mRNA | AK223482.1 | - | 9 - 10 | 1506 | 66 | |
| AK303249.1 | - | 8 - 9 | 1604 | 66 | ||
| AK314791.1 | - | 9 - 10 | 1676 | 66 | ||
| BC007585.1 | - | 9 - 10 | 1696 | 3162656 | 66 | |
| BC053555.1 | - | 9 - 10 | 1665 | 6140320 | 66 | |
| X13293.1 | - | 9 - 10 | 1633 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000320.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
regulation of cell cycle
mitotic spindle assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription factor activity, sequence-specific DNA binding
protein binding
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
regulation of cell cycle
mitotic spindle assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004580.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004580.hg.1 | ENST00000396863.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004580.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004580.hg.1 | TC20000320.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | TC2000007438.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000151539.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000217026.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | ENST00000396863.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_001278610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007438.hg.1 | NM_002466.3 | Refseq |

MYBL2
Genomic Map