Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001537.3 | NP_001528.1 | 1 - 2 | 219 | 103 | |
| GenBank mRNA | AF068754.1 | - | 1 - 2 | 99 | 103 | |
| AK172732.1 | - | 1 - 2 | 84 | 103 | ||
| BC007515.2 | - | 1 - 2 | 91 | 3010737 | 103 | |
| BX537440.1 | - | 1 - 2 | 106 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000650.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011714.hg.1 | ENST00000513509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011590.hg.1 | ENST00000299671.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009158.hg.1 | ENST00000433866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009158.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009158.hg.1 | TC16000650.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007311.hg.1 | ENST00000555205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009159.hg.1 | ENST00000433866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009159.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009159.hg.1 | TC16000650.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009160.hg.1 | ENST00000433866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009160.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009160.hg.1 | TC16000650.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009157.hg.1 | ENST00000433866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009157.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16009157.hg.1 | TC16000650.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164352.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011590.hg.1 | ENST00000299671.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164345.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164348.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164346.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011714.hg.1 | ENST00000513509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000569301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008627.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000567382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | ENST00000570259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164350.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600164343.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164343.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164343.hg.1 | ENST00000433866.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600164343.hg.1 | NM_001537.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600164343.hg.1 | TC1600008627.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007311.hg.1 | ENST00000555205.1 | Ensembl |

HSBP1
Genomic Map