Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK128540.1 | - | 3 - 4 | 432 | 184 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128540.1 transcripts.
Set Membership: |
|

HSPA13
Genomic Map