Assay Details
| Gene Symbol: HIVEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006734.3 | NP_006725.3 | 5 - 6 | 5928 | 87 | |
| XM_017010805.1 | XP_016866294.1 | 5 - 6 | 5893 | 87 | ||
| GenBank mRNA | AK226163.1 | - | 1 - 2 | 3406 | 87 | |
| AK296531.1 | - | 1 - 2 | 176 | 87 | ||
| CR936782.1 | - | 5 - 6 | 5928 | 87 | ||
| DQ231041.1 | - | 1 - 2 | 5185 | 87 | ||
| X65644.1 | - | 1 - 2 | 5362 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002181.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
multicellular organism development
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
multicellular organism development
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06027360.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027360.hg.1 | ENST00000012134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027360.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027360.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027360.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027359.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027359.hg.1 | ENST00000012134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027359.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027359.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027359.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027361.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027361.hg.1 | ENST00000012134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027361.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027361.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027361.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027362.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027362.hg.1 | ENST00000012134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027362.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027362.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027362.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027364.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027364.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027364.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027364.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027364.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | ENST00000012134.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | ENST00000367604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027365.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027366.hg.1 | TC06002181.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027366.hg.1 | ENST00000367603.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06027366.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192631.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192634.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192629.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192635.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192635.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192635.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192635.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192635.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013409.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192630.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000012134.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192628.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000367603.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000367604.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | ENST00000474532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | NM_006734.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600192633.hg.1 | TC0600013409.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 210854 | - | - |

HIVEP2
Genomic Map