Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007440.hg.1 |
TC0X000566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007440.hg.1 |
ENST00000371921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007440.hg.1 |
ENST00000451986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007440.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007442.hg.1 |
TC0X000566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007442.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007442.hg.1 |
ENST00000371921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007442.hg.1 |
ENST00000451986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007442.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006968.hg.1 |
ENST00000415420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007443.hg.1 |
TC0X000566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007443.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007443.hg.1 |
ENST00000371921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007443.hg.1 |
ENST00000451986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X007443.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155860.hg.1 |
TC0X00008195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008195.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155856.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155856.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155856.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155856.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155856.hg.1 |
TC0X00008195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
ENST00000371920.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
ENST00000371921.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
NM_016383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00155859.hg.1 |
TC0X00008195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000006968.hg.1 |
ENST00000415420.1 |
Ensembl |