Assay Details
| Gene Symbol: CCL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006664.3 | NP_006655.1 | 1 - 2 | 128 | 65 | |
| GenBank mRNA | AB010445.1 | - | 1 - 2 | 128 | 65 | |
| AF082393.1 | - | 1 - 2 | 69 | 65 | ||
| AJ243542.1 | - | 1 - 2 | 93 | 65 | ||
| BC148263.1 | - | 1 - 2 | 91 | 8860203 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09002914.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09013699.hg.1 | TC09002914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013699.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013699.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013700.hg.1 | TC09002914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013700.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013700.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013697.hg.1 | TC09002914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013697.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013697.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013697.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000416454.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000421828.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000423809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000544078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000555007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009914.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000557161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167982.hg.1 | TC0900009914.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167986.hg.1 | TC0900009914.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | ENST00000259631.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | ENST00000564224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | NM_006664.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167984.hg.1 | TC0900009914.hg.1 | Clariom D TC |

CCL27
Genomic Map