Assay Details
| Gene Symbol: GSTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000852.3 | NP_000843.1 | 2 - 3 | 285 | 70 | |
| GenBank mRNA | BC010915.1 | - | 2 - 3 | 72 | 3960700 | 70 |
| BT019949.1 | - | 1 - 2 | 36 | 70 | ||
| BT019950.1 | - | 1 - 2 | 36 | 70 | ||
| CA946272.1 | - | 2 - 3 | 285 | 70 | ||
| CR450361.1 | - | 1 - 2 | 36 | 70 | ||
| EU794620.1 | - | 2 - 3 | 65 | 70 | ||
| M69113.1 | - | 92 | 90 | |||
| U30897.1 | - | 2 - 3 | 48 | 70 | ||
| U62589.1 | - | 2 - 3 | 49 | 70 | ||
| X06547.1 | - | 1 - 2 | 42 | 70 | ||
| X15480.1 | - | 2 - 3 | 53 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000694.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
negative regulation of acute inflammatory response
negative regulation of protein kinase activity
glutathione metabolic process
xenobiotic metabolic process
central nervous system development
negative regulation of biosynthetic process
negative regulation of tumor necrosis factor-mediated signaling pathway
oligodendrocyte development
organ regeneration
response to estradiol
negative regulation of interleukin-1 beta production
negative regulation of tumor necrosis factor production
cellular response to insulin stimulus
regulation of stress-activated MAPK cascade
negative regulation of stress-activated MAPK cascade
positive regulation of superoxide anion generation
response to L-ascorbic acid
common myeloid progenitor cell proliferation
nitric oxide storage
negative regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
negative regulation of JUN kinase activity
linoleic acid metabolic process
response to ethanol
response to mercury ion
negative regulation of fibroblast proliferation
negative regulation of nitric-oxide synthase biosynthetic process
regulation of ERK1 and ERK2 cascade
negative regulation of ERK1 and ERK2 cascade
negative regulation of leukocyte proliferation
cellular response to lipopolysaccharide
cellular response to epidermal growth factor stimulus
cellular response to glucocorticoid stimulus
cellular response to cell-matrix adhesion
negative regulation of monocyte chemotactic protein-1 production
cellular oxidant detoxification
glutathione derivative biosynthetic process
negative regulation of extrinsic apoptotic signaling pathway
glutathione transferase activity
glutathione peroxidase activity
protein binding
drug binding
JUN kinase binding
kinase regulator activity
S-nitrosoglutathione binding
dinitrosyl-iron complex binding
glutathione binding
nitric oxide binding
response to reactive oxygen species
negative regulation of acute inflammatory response
negative regulation of protein kinase activity
glutathione metabolic process
xenobiotic metabolic process
central nervous system development
negative regulation of biosynthetic process
negative regulation of tumor necrosis factor-mediated signaling pathway
oligodendrocyte development
organ regeneration
response to estradiol
negative regulation of interleukin-1 beta production
negative regulation of tumor necrosis factor production
cellular response to insulin stimulus
regulation of stress-activated MAPK cascade
negative regulation of stress-activated MAPK cascade
positive regulation of superoxide anion generation
response to L-ascorbic acid
common myeloid progenitor cell proliferation
nitric oxide storage
negative regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
negative regulation of JUN kinase activity
linoleic acid metabolic process
response to ethanol
response to mercury ion
negative regulation of fibroblast proliferation
negative regulation of nitric-oxide synthase biosynthetic process
regulation of ERK1 and ERK2 cascade
negative regulation of ERK1 and ERK2 cascade
negative regulation of leukocyte proliferation
cellular response to lipopolysaccharide
cellular response to epidermal growth factor stimulus
cellular response to glucocorticoid stimulus
cellular response to cell-matrix adhesion
negative regulation of monocyte chemotactic protein-1 production
cellular oxidant detoxification
glutathione derivative biosynthetic process
negative regulation of extrinsic apoptotic signaling pathway
glutathione transferase activity
glutathione peroxidase activity
protein binding
drug binding
JUN kinase binding
kinase regulator activity
S-nitrosoglutathione binding
dinitrosyl-iron complex binding
glutathione binding
nitric oxide binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11009025.hg.1 | ENST00000398603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009025.hg.1 | ENST00000398606.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009025.hg.1 | ENST00000498765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009025.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11009025.hg.1 | TC11000694.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000467591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000495996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008144.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000398603.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000398606.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000489040.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000494593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | ENST00000498765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | NM_000852.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100159997.hg.1 | TC1100008144.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 214807 | - | - |

GSTP1
Genomic Map