Assay Details
| Gene Symbol: LIPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005357.3 | NP_005348.2 | 7 - 8 | 2645 | 74 | |
| XM_005258938.4 | XP_005258995.1 | 7 - 8 | 1631 | 74 | ||
| XM_005258939.3 | XP_005258996.2 | 8 - 9 | 1669 | 74 | ||
| XM_005258940.3 | XP_005258997.1 | 7 - 8 | 1570 | 74 | ||
| XM_006723218.2 | XP_006723281.1 | 7 - 8 | 1770 | 74 | ||
| XM_017026810.1 | XP_016882299.1 | 7 - 8 | 1510 | 74 | ||
| GenBank mRNA | AK292482.1 | - | 7 - 8 | 2625 | 74 | |
| BC070041.1 | - | 7 - 8 | 2645 | 5296155 | 74 | |
| U40002.1 | - | 7 - 8 | 2645 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001573.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
protein kinase binding
hormone-sensitive lipase activity
protein phosphorylation
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
protein kinase binding
hormone-sensitive lipase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021443.hg.1 | ENST00000244289.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021443.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021443.hg.1 | TC19001573.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | TC1900010802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189396.hg.1 | NM_005357.3 | Refseq |

LIPE
Genomic Map