Assay Details
| Gene Symbol: EXOC3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138568.3 | NP_612635.3 | 4 - 5 | 337 | 65 | |
| GenBank mRNA | AK093466.1 | - | 4 - 5 | 337 | 65 | |
| AK225791.1 | - | 3 - 4 | 335 | 65 | ||
| BC104947.1 | - | 3 - 4 | 323 | 8143950 | 65 | |
| BC104949.1 | - | 3 - 4 | 323 | 8143952 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001616.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19022053.hg.1 | ENST00000252482.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022053.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022053.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022053.hg.1 | TC19001616.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022054.hg.1 | ENST00000252482.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022054.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022054.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022054.hg.1 | TC19001616.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022052.hg.1 | ENST00000252482.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022052.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022052.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022052.hg.1 | TC19001616.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022051.hg.1 | ENST00000252482.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022051.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022051.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022051.hg.1 | TC19001616.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022055.hg.1 | ENST00000252482.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022055.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022055.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022055.hg.1 | TC19001616.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | TC1900010931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190679.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | TC1900010931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190680.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | TC1900010931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190681.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | TC1900010931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190682.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010931.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | NM_138568.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | TC1900010931.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | ENST00000252482.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | ENST00000413988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190678.hg.1 | NM_138568.4 | Refseq |

EXOC3L2
Genomic Map