Assay Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032288.6 | NP_115664.2 | 1 - 2 | 324 | 78 | |
| GenBank mRNA | AJ344094.1 | - | 1 - 2 | 170 | 78 | |
| AK027672.1 | - | 1 - 2 | 208 | 78 | ||
| AK296283.1 | - | 1 - 2 | 139 | 78 | ||
| AK315019.1 | - | 1 - 2 | 173 | 78 | ||
| AL136558.1 | - | 1 - 2 | 177 | 78 | ||
| BC035006.2 | - | 1 - 2 | 327 | 5267824 | 78 | |
| BC039734.1 | - | 1 - 2 | 234 | 6042926 | 78 | |
| BX509179.1 | - | 1 - 2 | 174 | 78 | ||
| CR533502.1 | - | 1 - 2 | 105 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001109.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000241502.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000412924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000415708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000418169.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000424384.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000426031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000426031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000426031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000426031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000428738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000492360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000492360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000492360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000492360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | ENST00000494309.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_001011537.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NM_032288.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010066.hg.1 | NR_027840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016456.hg.1 | ENST00000423946.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011030.hg.1 | ENST00000424250.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011030.hg.1 | ENST00000424250.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011030.hg.1 | ENST00000424250.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011030.hg.1 | ENST00000424250.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016456.hg.1 | ENST00000423946.2 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 209660 | - | - |

FYTTD1
Genomic Map