| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011259.hg.1 |
TC03000624.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011259.hg.1 |
ENST00000273668.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011259.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011259.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160732.hg.1 |
TC0300008536.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000273668.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000451944.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000465664.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490434.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
ENST00000490477.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008536.hg.1 |
NM_018456.5 |
Refseq |