Assay Details
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153605.3 | NP_705833.3 | 12 - 13 | 7864 | 99 | |
XM_005247117.4 | XP_005247174.1 | 11 - 12 | 7567 | 99 | ||
GenBank mRNA | AK091199.1 | - | 4 - 5 | 335 | 99 | |
BX647079.1 | - | 4 - 5 | 352 | 99 | ||
BX648353.1 | - | 9 - 10 | 1914 | 99 | ||
CR749313.1 | - | 9 - 10 | 989 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157671.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157671.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157671.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157671.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157671.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157674.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157674.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157674.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157674.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157674.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157675.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157675.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157675.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157675.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157675.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157676.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157676.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157676.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157676.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157676.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157672.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157672.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157672.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157672.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157672.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157679.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157679.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157679.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157679.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157679.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000485253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | ENST00000495403.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008088.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157677.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157677.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157677.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157677.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157677.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157670.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157670.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157670.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157670.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157670.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300157673.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157673.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157673.hg.1 | ENST00000389622.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300157673.hg.1 | NM_153605.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300157673.hg.1 | TC0300008088.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 105842 | - | - |
