Assay Details
| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198505.2 | NP_940907.2 | 20 - 21 | 2446 | 123 | |
| XM_011512770.1 | XP_011511072.1 | 20 - 21 | 2545 | 123 | ||
| XM_017006304.1 | XP_016861793.1 | 16 - 17 | 1759 | 123 | ||
| XM_017006305.1 | XP_016861794.1 | 7 - 8 | 862 | 123 | ||
| GenBank mRNA | AY823163.1 | - | 20 - 21 | 2446 | 123 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002120.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03037419.hg.1 | TC03002120.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037419.hg.1 | ENST00000342358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037419.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037419.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037420.hg.1 | TC03002120.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037420.hg.1 | ENST00000342358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037420.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037420.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201102.hg.1 | TC0300013563.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201101.hg.1 | TC0300013563.hg.1 | Clariom D TC |

ATP13A5
Genomic Map