Assay Details
| Gene Symbol: ATP13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198505.2 | NP_940907.2 | 12 - 13 | 1458 | 67 | |
| XM_011512770.1 | XP_011511072.1 | 12 - 13 | 1557 | 67 | ||
| XM_017006304.1 | XP_016861793.1 | 8 - 9 | 771 | 67 | ||
| GenBank mRNA | AK122613.1 | - | 1578 | 67 | ||
| AY823163.1 | - | 12 - 13 | 1458 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002120.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03037431.hg.1 | TC03002120.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037431.hg.1 | ENST00000342358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037431.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000446087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000488957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000495496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | ENST00000620313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013563.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201113.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201113.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201113.hg.1 | ENST00000342358.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201113.hg.1 | NM_198505.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201113.hg.1 | TC0300013563.hg.1 | Clariom D TC |

ATP13A5
Genomic Map