Assay Details
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127603.1 | NP_001121075.1 | 2 - 3 | 276 | 112 | |
NM_017881.2 | NP_060351.1 | 2 - 3 | 276 | 112 | ||
GenBank mRNA | AK000566.1 | - | 2 - 3 | 241 | 112 | |
AK023413.1 | - | 2 - 3 | 268 | 112 | ||
BC036804.1 | - | 2 - 3 | 276 | 5727308 | 112 | |
CB135264.1 | - | 2 - 3 | 276 | 112 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001220.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK000566.1,AK023413.1,BC036804.1,NM_001127603.1,NM_017881.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | TC09001220.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | ENST00000361092.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | ENST00000376794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | ENST00000376808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | ENST00000376811.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09015649.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000466137.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000482537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000490321.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010445.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000361092.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000376808.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000376811.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | ENST00000494066.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NM_001127603.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NM_017881.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | NR_023352.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900171479.hg.1 | TC0900010445.hg.1 | Clariom D TC |
