Assay Details
| Gene Symbol: DDIAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058145.1 | - | 8 | 1637 | 117 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream |
RAB30, member RAS oncogene family |
RAB30 | 14.9kb | Anti-sense | Chr. 11 - 82973133 - 83071923 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DDIAS
Genomic Map