| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003635.hg.1 |
ENST00000263379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003635.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003635.hg.1 |
TC19000250.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011677.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199588.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199588.hg.1 |
NM_004843.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199588.hg.1 |
TC1900011677.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199588.hg.1 |
ENST00000263379.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199588.hg.1 |
NM_004843.3 |
Refseq |