Assay Details
| Gene Symbol: MKRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014160.4 | NP_054879.3 | 2 - 3 | 294 | 95 | |
| GenBank mRNA | AF302084.1 | - | 2 - 3 | 208 | 95 | |
| AK091318.1 | - | 2 - 3 | 239 | 95 | ||
| BC015715.1 | - | 2 - 3 | 236 | 3916199 | 95 | |
| DR157266.1 | - | 2 - 3 | 294 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000072.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03001615.hg.1 | TC03000072.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001615.hg.1 | ENST00000170447.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001615.hg.1 | ENST00000411987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001615.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001615.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146128.hg.1 | TC0300006631.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300146127.hg.1 | TC0300006631.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000170447.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000411987.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | ENST00000448482.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_001271707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006631.hg.1 | NM_014160.4 | Refseq |

MKRN2
Genomic Map