Assay Details
| Gene Symbol: USP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290325.1 | NP_001277254.1 | 8 - 9 | 2135 | 92 | |
| NM_001290326.1 | NP_001277255.1 | 9 - 10 | 2251 | 92 | ||
| NM_032557.6 | NP_115946.2 | 8 - 9 | 2135 | 92 | ||
| XM_011532360.2 | XP_011530662.1 | 7 - 8 | 1521 | 92 | ||
| GenBank mRNA | AB067478.1 | - | 7 - 8 | 816 | 92 | |
| AK057992.1 | - | 5 - 6 | 659 | 92 | ||
| AK093754.1 | - | 9 - 10 | 1474 | 92 | ||
| AK126943.1 | - | 9 - 10 | 1963 | 92 | ||
| AL833976.1 | - | 8 - 9 | 2107 | 92 | ||
| BC039115.1 | - | 8 - 9 | 2135 | 5258804 | 92 | |
| BC068975.1 | - | 8 - 9 | 2082 | 6067501 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000696.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009887.hg.1 | TC04000696.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009887.hg.1 | ENST00000307017.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009887.hg.1 | ENST00000510377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04009887.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160887.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160887.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160887.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160887.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160887.hg.1 | TC0400008876.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008876.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000307017.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000510377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | ENST00000511739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_001290325.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_001290326.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | NM_032557.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400160886.hg.1 | TC0400008876.hg.1 | Clariom D TC |

USP38
Genomic Map