Assay Details
| Gene Symbol: YEATS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018023.4 | NP_060493.3 | 26 - 27 | 3779 | 99 | |
| XM_005247597.2 | XP_005247654.1 | 26 - 27 | 3796 | 99 | ||
| XM_005247598.4 | XP_005247655.1 | 26 - 27 | 3704 | 99 | ||
| XM_006713688.3 | XP_006713751.1 | 26 - 27 | 3914 | 99 | ||
| XM_011512965.1 | XP_011511267.1 | 25 - 26 | 3634 | 99 | ||
| XM_017006809.1 | XP_016862298.1 | 26 - 27 | 3910 | 99 | ||
| XM_017006810.1 | XP_016862299.1 | 26 - 27 | 3701 | 99 | ||
| XM_017006811.1 | XP_016862300.1 | 25 - 26 | 3566 | 99 | ||
| XM_017006813.1 | XP_016862302.1 | 25 - 26 | 3600 | 99 | ||
| XM_017006814.1 | XP_016862303.1 | 25 - 26 | 3581 | 99 | ||
| GenBank mRNA | AB033023.1 | - | 26 - 27 | 3779 | 99 | |
| AK023370.1 | - | 12 - 13 | 1307 | 99 | ||
| BC043393.1 | - | 12 - 13 | 1730 | 99 | ||
| BC082270.1 | - | 9 - 10 | 1084 | 99 | ||
| BC150273.1 | - | 26 - 27 | 3779 | 9007245 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000986.hg.1
Set Membership: |
|

YEATS2
Genomic Map