Assay Details
| Gene Symbol: YEATS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018023.4 | NP_060493.3 | 9 - 10 | 1161 | 62 | |
| XM_005247597.2 | XP_005247654.1 | 9 - 10 | 1175 | 62 | ||
| XM_005247598.4 | XP_005247655.1 | 9 - 10 | 1083 | 62 | ||
| XM_006713688.3 | XP_006713751.1 | 9 - 10 | 1293 | 62 | ||
| XM_011512965.1 | XP_011511267.1 | 8 - 9 | 1013 | 62 | ||
| XM_011512966.1 | XP_011511268.1 | 9 - 10 | 1175 | 62 | ||
| XM_017006809.1 | XP_016862298.1 | 9 - 10 | 1292 | 62 | ||
| XM_017006810.1 | XP_016862299.1 | 9 - 10 | 1083 | 62 | ||
| XM_017006811.1 | XP_016862300.1 | 8 - 9 | 945 | 62 | ||
| XM_017006812.1 | XP_016862301.1 | 9 - 10 | 1182 | 62 | ||
| XM_017006814.1 | XP_016862303.1 | 8 - 9 | 960 | 62 | ||
| GenBank mRNA | AB033023.1 | - | 9 - 10 | 1161 | 62 | |
| BC150273.1 | - | 9 - 10 | 1161 | 9007245 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000986.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017006814.1 transcripts.
Set Membership: |
|

YEATS2
Genomic Map