Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 19 - 20 | 2614 | 109 | |
| NM_001204516.1 | NP_001191445.1 | 17 - 18 | 2318 | 109 | ||
| NM_001204517.1 | NP_001191446.1 | 18 - 19 | 3559 | 109 | ||
| NM_014970.3 | NP_055785.2 | 18 - 19 | 2450 | 109 | ||
| XM_005244970.1 | XP_005245027.1 | 18 - 19 | 2453 | 109 | ||
| XM_011509307.2 | XP_011507609.2 | 18 - 19 | 2453 | 109 | ||
| GenBank mRNA | AK026471.1 | - | 18 - 19 | 2229 | 109 | |
| AK122897.1 | - | 18 - 19 | 3561 | 109 | ||
| AK297368.1 | - | 17 - 18 | 2245 | 109 | ||
| AK301912.1 | - | 11 - 12 | 1417 | 109 | ||
| AK315586.1 | - | 18 - 19 | 2364 | 109 | ||
| BC028679.2 | - | 18 - 19 | 2407 | 4829354 | 109 | |
| BC048012.1 | - | 17 - 18 | 2362 | 109 | ||
| U59919.1 | - | 18 - 19 | 2367 | 109 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map