Assay Details
| Gene Symbol: KIF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004523.3 | NP_004514.2 | 10 - 11 | 1523 | 128 | |
| GenBank mRNA | AK314258.1 | - | 10 - 11 | 1426 | 128 | |
| BC126211.1 | - | 10 - 11 | 1322 | 8991927 | 128 | |
| BC136474.1 | - | 10 - 11 | 1322 | 9020463 | 128 | |
| U37426.1 | - | 10 - 11 | 1305 | 128 | ||
| X85137.1 | - | 10 - 11 | 1225 | 128 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000659.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
spindle organization
mitotic spindle organization
chromosome segregation
mitotic nuclear division
mitotic centrosome separation
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of mitotic centrosome separation
cell division
mitotic spindle assembly
microtubule motor activity
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
protein kinase binding
protein complex binding
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
spindle organization
mitotic spindle organization
chromosome segregation
mitotic nuclear division
mitotic centrosome separation
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of mitotic centrosome separation
cell division
mitotic spindle assembly
microtubule motor activity
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
protein kinase binding
protein complex binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10007382.hg.1 | ENST00000260731.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007382.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007382.hg.1 | TC10000659.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008471.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157521.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157521.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157521.hg.1 | ENST00000260731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157521.hg.1 | NM_004523.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157521.hg.1 | TC1000008471.hg.1 | Clariom D TC |

KIF11
Genomic Map