Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024307.2 | NP_077283.2 | 3 - 4 | 396 | 70 | |
| GenBank mRNA | AK026256.1 | - | 2 - 3 | 604 | 70 | |
| AK225450.1 | - | 2 - 3 | 604 | 70 | ||
| BC002714.2 | - | 3 - 4 | 396 | 3629566 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001020.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16014589.hg.1 | ENST00000360688.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014589.hg.1 | ENST00000406256.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014589.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014589.hg.1 | TC16001020.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600176762.hg.1 | TC1600009953.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000360688.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000406256.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000565704.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566434.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | ENST00000566613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009953.hg.1 | NM_024307.2 | Refseq |

GDPD3
Genomic Map