Assay Details
| Gene Symbol: CELSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014246.1 | NP_055061.1 | 17 - 18 | 6343 | 77 | |
| XM_006724383.3 | XP_006724446.1 | 17 - 18 | 6670 | 77 | ||
| XM_011530554.2 | XP_011528856.1 | 17 - 18 | 2886 | 77 | ||
| XM_011530555.2 | XP_011528857.1 | 17 - 18 | 2967 | 77 | ||
| GenBank mRNA | AF231024.1 | - | 17 - 18 | 6343 | 77 | |
| AK302517.1 | - | 9 - 10 | 1421 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000889.hg.1
Set Membership: |
|

CELSR1
Genomic Map