Assay Details
| Gene Symbol: TSGA10IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152762.2 | NP_689975.2 | 1 - 2 | 177 | 71 | |
| XM_011544887.2 | XP_011543189.1 | 1 - 2 | 376 | 71 | ||
| GenBank mRNA | AK057442.1 | - | 1 - 2 | 177 | 71 | |
| BC104022.1 | - | 1 - 2 | 177 | 40025091 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000648.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | ENST00000528291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | ENST00000534740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008146.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008150.hg.1 | ENST00000528291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008150.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008150.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008150.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008150.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008145.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008145.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008145.hg.1 | ENST00000534740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008145.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008145.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | ENST00000528291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | ENST00000534740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008148.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008151.hg.1 | ENST00000528291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008151.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008151.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008151.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008151.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008152.hg.1 | ENST00000532620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008152.hg.1 | ENST00000533763.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008152.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11008152.hg.1 | TC11000648.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158600.hg.1 | TC1100008045.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158596.hg.1 | TC1100008045.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158602.hg.1 | TC1100008045.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158598.hg.1 | TC1100008045.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000531518.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000534740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100008045.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000528291.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000532620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | ENST00000608857.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | NM_152762.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100158601.hg.1 | TC1100008045.hg.1 | Clariom D TC |

TSGA10IP
Genomic Map