Assay Details
| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056654.1 | - | 1 | 631 | 116 | |
| AK308756.1 | - | 6 | 116 | |||
| Gene Symbol: LOC100129034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027406.1 | 1 | 3008 | 116 | ||
| GenBank mRNA | AK090539.1 | - | 1 | 2212 | 116 | |
| CR627469.1 | - | 1 | 784 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P91042 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 16768 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10904 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P91042 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P91042 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955681 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27301147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27301147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262193 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955686 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955682 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955673 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955684 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955676 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06955681 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955681 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262202 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955682 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955699 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955684 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262200 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262198 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262196 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955712 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955675 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955694 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955701 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262193 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06955687 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16262191 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 244823_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 225214_PM_at | - | - |
| Affymetrix Human Clariom D Assay | PSR0900158179.hg.1 | NR_027406.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158179.hg.1 | NR_027406.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158179.hg.1 | TC0900008726.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158180.hg.1 | NR_027406.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158180.hg.1 | NR_027406.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158180.hg.1 | TC0900008726.hg.1 | Clariom D TC |
| Human Genome U133 Set | 244823_PM_at | - | - |
| Human Genome U133 Set | 244823_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039224 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039224 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 170281 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 231958 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 232290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3256478 | NR_027406.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3256478 | AK056654.1 | human_transcriptome |

PSMB7
Genomic Map