Assay Details
| Gene Symbol: XRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282857.1 | NP_001269786.1 | 15 - 16 | 1831 | 68 | |
| NM_019001.4 | NP_061874.3 | 15 - 16 | 1831 | 68 | ||
| XM_006713673.2 | XP_006713736.1 | 15 - 16 | 1831 | 68 | ||
| XM_011512919.2 | XP_011511221.1 | 15 - 16 | 1831 | 68 | ||
| XM_011512920.2 | XP_011511222.1 | 15 - 16 | 1831 | 68 | ||
| XM_011512922.2 | XP_011511224.1 | 12 - 13 | 1462 | 68 | ||
| XM_017006640.1 | XP_016862129.1 | 15 - 16 | 1831 | 68 | ||
| XM_017006641.1 | XP_016862130.1 | 15 - 16 | 1831 | 68 | ||
| XM_017006642.1 | XP_016862131.1 | 15 - 16 | 1831 | 68 | ||
| GenBank mRNA | AK123897.1 | - | 12 - 13 | 1304 | 68 | |
| AY137776.1 | - | 15 - 16 | 1713 | 68 | ||
| BC078146.1 | - | 15 - 16 | 1743 | 68 | ||
| BX640905.1 | - | 15 - 16 | 1780 | 68 | ||
| CR627396.1 | - | 15 - 16 | 1783 | 68 | ||
| CR749518.1 | - | 15 - 16 | 1761 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001851.hg.1
Set Membership: |
|

XRN1
Genomic Map