Assay Details
| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017008864.1 | XP_016864353.1 | 2 - 3 | 150 | 68 | |
| XM_017008865.1 | XP_016864354.1 | 2 - 3 | 3266 | 68 | ||
| XM_017008866.1 | XP_016864355.1 | 2 - 3 | 340 | 68 | ||
| XM_017008867.1 | XP_016864356.1 | 2 - 3 | 146 | 68 | ||
| XM_017008868.1 | XP_016864357.1 | 2 - 3 | 340 | 68 | ||
| GenBank mRNA | BC001279.1 | - | 2 - 3 | 479 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC001279.1, XM_017008864.1, XM_017008865.1, XM_017008866.1, XM_017008867.1, XM_017008868.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |

MFAP3L
Genomic Map