Assay Details
| Gene Symbol: GBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319179.1 | NP_001306108.1 | 1 - 2 | 183 | 99 | |
| NM_001319180.1 | NP_001306109.1 | 1 - 2 | 183 | 99 | ||
| NM_001319181.1 | NP_001306110.1 | 1 - 2 | 183 | 99 | ||
| NM_018284.2 | NP_060754.2 | 1 - 2 | 183 | 99 | ||
| NR_135007.1 | 1 - 2 | 183 | 99 | |||
| XM_005270743.2 | XP_005270800.1 | 1 - 2 | 177 | 99 | ||
| XM_006710574.2 | XP_006710637.1 | 1 - 2 | 197 | 99 | ||
| XM_011541234.1 | XP_011539536.1 | 1 - 2 | 197 | 99 | ||
| GenBank mRNA | AK001823.1 | - | 1 - 2 | 155 | 99 | |
| AK309096.1 | - | 1 - 2 | 168 | 99 | ||
| AL136680.1 | - | 1 - 2 | 178 | 99 | ||
| BC017992.1 | - | 1 - 2 | 155 | 99 | ||
| BC063819.1 | - | 1 - 2 | 95 | 99 | ||
| BC140837.1 | - | 1 - 2 | 68 | 9021705 | 99 | |
| BC144650.1 | - | 1 - 2 | 68 | 99 | ||
| BP281050.1 | - | 1 - 2 | 183 | 99 | ||
| CR936755.1 | - | 1 - 2 | 184 | 99 | ||
| CR936758.1 | - | 1 - 2 | 124 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AL136680.1, BC017992.1, BC140837.1, NM_001319179.1, NM_001319180.1, NM_001319181.1, NM_018284.2, XM_005270743.2, XM_006710574.2, XM_011541234.1 transcripts.
Set Membership: |
|

GBP3
Genomic Map