Assay Details
| Gene Symbol: LINC01534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110707.1 | 1 - 2 | 179 | 114 | ||
| NR_110708.1 | 1 - 2 | 179 | 114 | |||
| GenBank mRNA | BM701845.1 | - | 1 - 2 | 180 | 114 | |
| BQ049787.1 | - | 1 - 2 | 140 | 114 | ||
| DA064533.1 | - | 1 - 2 | 164 | 114 | ||
| DA195154.1 | - | 1 - 2 | 164 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | TC1900010532.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186568.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | TC1900010532.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186569.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | TC1900010532.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900186570.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000425254.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000433232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | ENST00000590952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110707.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010532.hg.1 | NR_110708.1 | Refseq |

LINC01534
Genomic Map