Assay Details
| Gene Symbol: LY86-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091027.1 | - | 6 - 7 | 1052 | 140 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000403621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000403621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000429345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000435641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000447858.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000606992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | ENST00000607278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600010712.hg.1 | NR_026970.1 | Refseq |

LY86-AS1
Genomic Map