Assay Details
| Gene Symbol: AGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006408.3 | NP_006399.1 | 1 - 2 | 181 | 78 | |
| GenBank mRNA | AF038451.1 | - | 1 - 2 | 52 | 78 | |
| AF087879.1 | - | 1 - 2 | 143 | 78 | ||
| AF088867.1 | - | 1 - 2 | 141 | 78 | ||
| AF115926.1 | - | 1 - 2 | 51 | 78 | ||
| AK223067.1 | - | 1 - 2 | 72 | 78 | ||
| AK296721.1 | - | 1 - 2 | 62 | 78 | ||
| AK312942.1 | - | 1 - 2 | 62 | 78 | ||
| AY359009.1 | - | 1 - 2 | 141 | 78 | ||
| BC015503.1 | - | 1 - 2 | 50 | 3852448 | 78 | |
| BF680655.1 | - | 1 - 2 | 182 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001163.hg
Additional Information:
This assay is in the 5'UTR region of the AF038451.1,AF087879.1,AF088867.1,AF115926.1,AK296721.1,AK312942.1,AY359009.1,BC015503.1,NM_006408.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of gene expression
positive regulation of cell-substrate adhesion
response to endoplasmic reticulum stress
positive regulation of epidermal growth factor receptor signaling pathway
digestive tract morphogenesis
positive regulation of developmental growth
lung goblet cell differentiation
negative regulation of cell death
mucus secretion
positive regulation of establishment of protein localization to plasma membrane
positive regulation of IRE1-mediated unfolded protein response
positive regulation of PERK-mediated unfolded protein response
dystroglycan binding
epidermal growth factor receptor binding
protein binding
protein homodimerization activity
positive regulation of gene expression
positive regulation of cell-substrate adhesion
response to endoplasmic reticulum stress
positive regulation of epidermal growth factor receptor signaling pathway
digestive tract morphogenesis
positive regulation of developmental growth
lung goblet cell differentiation
negative regulation of cell death
mucus secretion
positive regulation of establishment of protein localization to plasma membrane
positive regulation of IRE1-mediated unfolded protein response
positive regulation of PERK-mediated unfolded protein response
dystroglycan binding
epidermal growth factor receptor binding
protein binding
protein homodimerization activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07018206.hg.1 | TC07001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018206.hg.1 | ENST00000401412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018206.hg.1 | ENST00000419304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018206.hg.1 | ENST00000419572.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018206.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018209.hg.1 | TC07001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018209.hg.1 | ENST00000401412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018209.hg.1 | ENST00000419304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018209.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018209.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018213.hg.1 | TC07001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018213.hg.1 | ENST00000419304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018213.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018214.hg.1 | TC07001163.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07018214.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173617.hg.1 | TC0700010355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700173625.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173625.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173625.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173625.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173625.hg.1 | TC0700010355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700173626.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173626.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173626.hg.1 | TC0700010355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000412973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000450569.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010355.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000401412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000419304.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000468419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000486219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | ENST00000489523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | NM_006408.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700173621.hg.1 | TC0700010355.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195456 | - | - |

AGR2
Genomic Map