Assay Details
| Gene Symbol: XPNPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003399.5 | NP_003390.4 | 6 - 7 | 741 | 62 | |
| GenBank mRNA | AK300954.1 | - | 6 - 7 | 762 | 62 | |
| BC126174.1 | - | 6 - 7 | 658 | 8991890 | 62 | |
| BC143901.1 | - | 6 - 7 | 658 | 9052419 | 62 | |
| U90724.2 | - | 6 - 7 | 761 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000617.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008108.hg.1 | TC0X000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008108.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008108.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371105.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008387.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157093.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157093.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157093.hg.1 | ENST00000371106.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00157093.hg.1 | NM_003399.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00157093.hg.1 | TC0X00008387.hg.1 | Clariom D TC |

XPNPEP2
Genomic Map