Assay Details
| Gene Symbol: SMOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034852.2 | NP_001030024.1 | 5 - 6 | 813 | 70 | |
| NM_022137.5 | NP_071420.1 | 5 - 6 | 813 | 70 | ||
| GenBank mRNA | AJ249900.1 | - | 5 - 6 | 785 | 70 | |
| AK289988.1 | - | 5 - 6 | 794 | 70 | ||
| AK313063.1 | - | 5 - 6 | 794 | 70 | ||
| BC008608.1 | - | 5 - 6 | 781 | 4158532 | 70 | |
| BC011548.1 | - | 5 - 6 | 784 | 4549051 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000419.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14005095.hg.1 | ENST00000361956.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005095.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005095.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005095.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005095.hg.1 | TC14000419.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000553839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000555917.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | ENST00000557483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007566.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | ENST00000361956.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | ENST00000381280.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | NM_001034852.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | NM_022137.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400153085.hg.1 | TC1400007566.hg.1 | Clariom D TC |

SMOC1
Genomic Map