Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018310.3 | NP_060780.2 | 1 - 2 | 281 | 68 | |
| GenBank mRNA | AF206673.2 | - | 1 - 2 | 281 | 68 | |
| AF298153.1 | - | 1 - 2 | 204 | 68 | ||
| AK001914.1 | - | 1 - 2 | 243 | 68 | ||
| AK294337.1 | - | 2 - 3 | 240 | 68 | ||
| AK315420.1 | - | 1 - 2 | 230 | 68 | ||
| BC010648.1 | - | 1 - 2 | 234 | 3871326 | 68 | |
| DA688249.1 | - | 1 - 2 | 281 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001137.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08014577.hg.1 | TC08001137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014577.hg.1 | ENST00000220659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014577.hg.1 | ENST00000520601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014577.hg.1 | ENST00000521170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014577.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | TC08001137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | ENST00000220659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | ENST00000520601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | ENST00000521170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | ENST00000545765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014578.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014582.hg.1 | TC08001137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014582.hg.1 | ENST00000220659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014582.hg.1 | ENST00000545765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014582.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | TC08001137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | ENST00000220659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | ENST00000520601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | ENST00000521170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | ENST00000545765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014575.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | TC08001137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | ENST00000220659.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | ENST00000520601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | ENST00000521170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | ENST00000545765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014576.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168906.hg.1 | TC0800010137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168908.hg.1 | TC0800010137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800168912.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168912.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168912.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168912.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168912.hg.1 | TC0800010137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800168907.hg.1 | TC0800010137.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000220659.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000520601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000521170.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | ENST00000522539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010137.hg.1 | NM_018310.3 | Refseq |

BRF2
Genomic Map