Assay Details
| Gene Symbol: COL9A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001853.3 | NP_001844.3 | 6 - 7 | 347 | 132 | |
| XM_011528545.1 | XP_011526847.1 | 6 - 7 | 345 | 132 | ||
| XM_017027666.1 | XP_016883155.1 | 6 - 7 | 345 | 132 | ||
| GenBank mRNA | BC011705.2 | - | 6 - 7 | 347 | 3830925 | 132 |
| L41162.1 | - | 6 - 7 | 344 | 132 | ||
More Information
Set Membership: |
|

COL9A3
Genomic Map