Assay Details
| Gene Symbol: SYT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020826.2 | NP_065877.1 | 1 - 2 | 309 | 83 | |
| GenBank mRNA | AB037848.1 | - | 1 - 2 | 295 | 83 | |
| AK291009.1 | - | 1 - 2 | 230 | 83 | ||
| AL512743.1 | - | 1 - 2 | 232 | 83 | ||
| BC093830.1 | - | 1 - 2 | 229 | 7939675 | 83 | |
| BC093832.1 | - | 1 - 2 | 231 | 7939677 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001585.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11019754.hg.1 | ENST00000020926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019754.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019754.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019754.hg.1 | TC11001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019759.hg.1 | ENST00000020926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019759.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019759.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019759.hg.1 | TC11001585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019757.hg.1 | ENST00000020926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019757.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019757.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019757.hg.1 | TC11001585.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180588.hg.1 | TC1100010654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180596.hg.1 | TC1100010654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180594.hg.1 | TC1100010654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180592.hg.1 | TC1100010654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100180593.hg.1 | TC1100010654.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000020926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000528101.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | ENST00000533332.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_001247987.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010654.hg.1 | NM_020826.2 | Refseq |

SYT13
Genomic Map