Assay Details
| Gene Symbol: CRYM-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC056679.1 | - | 1 - 2 | 218 | 103 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000338573.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000444326.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | ENST00000561968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007150.hg.1 | NR_026675.1 | Refseq |

CRYM-AS1
Genomic Map