Assay Details
| Gene Symbol: CA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005182.2 | NP_005173.1 | 1 - 2 | 147 | 78 | |
| XM_011523311.1 | XP_011521613.1 | 1 - 2 | 141 | 78 | ||
| GenBank mRNA | AY075019.1 | - | 1 - 2 | 154 | 78 | |
| BC033865.1 | - | 1 - 2 | 103 | 5185069 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000522.hg
Additional Information:
This assay is in the 5'UTR region of the AY075019.1,BC033865.1,NM_005182.2,XM_011523311.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
one-carbon metabolic process
bicarbonate transport
positive regulation of synaptic transmission, GABAergic
positive regulation of cellular pH reduction
mitophagy in response to mitochondrial depolarization
xenophagy
regulation of chloride transport
carbonate dehydratase activity
zinc ion binding
positive regulation of defense response to virus by host
one-carbon metabolic process
bicarbonate transport
positive regulation of synaptic transmission, GABAergic
positive regulation of cellular pH reduction
mitophagy in response to mitochondrial depolarization
xenophagy
regulation of chloride transport
carbonate dehydratase activity
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16007282.hg.1 | ENST00000338437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007282.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007282.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007282.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007282.hg.1 | TC16000522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007278.hg.1 | ENST00000338437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007278.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007278.hg.1 | TC16000522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007279.hg.1 | ENST00000338437.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007279.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16007279.hg.1 | TC16000522.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159960.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159960.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159960.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159960.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159960.hg.1 | TC1600008134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008134.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159959.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159959.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159959.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159959.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159959.hg.1 | TC1600008134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600159958.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159958.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159958.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159958.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159958.hg.1 | TC1600008134.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000338437.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000394069.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | ENST00000548332.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | NM_001014435.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | NM_005182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600159963.hg.1 | TC1600008134.hg.1 | Clariom D TC |

CA7
Genomic Map