Assay Details
| Gene Symbol: PID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100818.1 | NP_001094288.1 | 1 - 2 | 328 | 110 | |
| GenBank mRNA | AB075874.1 | - | 2 - 3 | 194 | 110 | |
| AK096636.1 | - | 1 - 2 | 270 | 110 | ||
| DB474424.1 | - | 1 - 2 | 328 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002845.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK096636.1,NM_001100818.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein phosphorylation
energy reserve metabolic process
positive regulation of gene expression
regulation of mitochondrial fusion
cellular response to leptin stimulus
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of insulin receptor signaling pathway
regulation of mitochondrial membrane potential
positive regulation of fat cell proliferation
mitochondrion morphogenesis
cellular response to cytokine stimulus
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to fatty acid
negative regulation of establishment of protein localization to plasma membrane
negative regulation of mitochondrial DNA replication
regulation of G1/S transition of mitotic cell cycle
regulation of reactive oxygen species metabolic process
positive regulation of reactive oxygen species metabolic process
negative regulation of ATP biosynthetic process
positive regulation of ATP biosynthetic process
negative regulation of glucose import in response to insulin stimulus
protein binding
negative regulation of protein phosphorylation
energy reserve metabolic process
positive regulation of gene expression
regulation of mitochondrial fusion
cellular response to leptin stimulus
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of insulin receptor signaling pathway
regulation of mitochondrial membrane potential
positive regulation of fat cell proliferation
mitochondrion morphogenesis
cellular response to cytokine stimulus
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to fatty acid
negative regulation of establishment of protein localization to plasma membrane
negative regulation of mitochondrial DNA replication
regulation of G1/S transition of mitotic cell cycle
regulation of reactive oxygen species metabolic process
positive regulation of reactive oxygen species metabolic process
negative regulation of ATP biosynthetic process
positive regulation of ATP biosynthetic process
negative regulation of glucose import in response to insulin stimulus
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | TC02002845.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | ENST00000392054.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | ENST00000392055.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | ENST00000482518.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | ENST00000542363.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045907.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | TC02002845.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | ENST00000392054.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | ENST00000392055.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045913.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045917.hg.1 | TC02002845.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045917.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045917.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015951.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217342.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217342.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217342.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217342.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217342.hg.1 | TC0200015951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000354069.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000482518.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217330.hg.1 | TC0200015951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217338.hg.1 | TC0200015951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217336.hg.1 | TC0200015951.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000392054.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000392055.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000409462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | ENST00000534952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | NM_001100818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | NM_017933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200217337.hg.1 | TC0200015951.hg.1 | Clariom D TC |

PID1
Genomic Map