Assay Details
| Gene Symbol: ATG2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720187.2 | XP_006720250.1 | 27 - 28 | 4350 | 71 | |
| XM_017021397.1 | XP_016876886.1 | 27 - 28 | 4353 | 71 | ||
| GenBank mRNA | AK128275.1 | - | 8 - 9 | 798 | 71 | |
| BC015016.2 | - | 1 - 2 | 81 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ATG2B
Genomic Map