Assay Details
| Gene Symbol: ATG2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720187.2 | XP_006720250.1 | 41 - 42 | 6363 | 61 | |
| GenBank mRNA | AK001104.1 | - | 13 - 14 | 1764 | 61 | |
| AK128275.1 | - | 22 - 23 | 2811 | 61 | ||
| BC015016.2 | - | 15 - 16 | 2094 | 61 | ||
More Information
Set Membership: |
|

ATG2B
Genomic Map