Assay Details
| Gene Symbol: LOC100287210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC009871.2 | - | 1 | 565 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | 5-hydroxytryptamine receptor 5A | HTR5A | 5-HT5A | -135.5kb | Sense | Chr. 7 - 155070324 - 155087392 on NCBI Build GRCh38 |
| Downstream | insulin induced gene 1 | INSIG1 |
CL-6, CL6 |
73.9kb | Sense | Chr. 7 - 155297776 - 155310235 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886566 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886522 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886562 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155340 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886598 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886541 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886604 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886587 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16044561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886571 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886549 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886547 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155330 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886594 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155342 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886589 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886595 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155321 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886573 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155378 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155323 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26155320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05886603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16044547 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009803.hg.1 | ENST00000624248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009803.hg.1 | ENST00000624248.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1555206_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1555206_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1555206_at7 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 201563 | - | - |

LOC100287210
Genomic Map