Assay Details
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004686.4 | NP_004677.3 | 6 - 7 | 966 | 77 | |
XM_011544692.2 | XP_011542994.1 | 3 - 4 | 405 | 77 | ||
GenBank mRNA | AF073482.1 | - | 2 - 3 | 235 | 77 | |
AK294468.1 | - | 6 - 7 | 727 | 77 | ||
AK316022.1 | - | 6 - 7 | 776 | 77 | ||
AK316283.1 | - | 6 - 7 | 786 | 77 | ||
BC130577.1 | - | 6 - 7 | 752 | 40146608 | 77 | |
BC130579.1 | - | 6 - 7 | 752 | 40146610 | 77 | |
CR749240.1 | - | 6 - 7 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001012.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
phosphatidylinositol-3-phosphatase activity
protein tyrosine phosphatase activity
protein binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
inositol phosphate dephosphorylation
phosphatidylinositol dephosphorylation
phosphatidylinositol-3-phosphatase activity
protein tyrosine phosphatase activity
protein binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08012702.hg.1 | TC08001012.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08012702.hg.1 | ENST00000180173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012702.hg.1 | ENST00000521857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08012702.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519590.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000519763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521177.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | ENST00000523571.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800009726.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000180173.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000517317.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000518272.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000519122.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | ENST00000521857.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | NM_004686.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800166095.hg.1 | TC0800009726.hg.1 | Clariom D TC |
