Assay Details
| Gene Symbol: BCAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003657.3 | NP_003648.2 | 7 - 8 | 1482 | 124 | |
| XM_005260587.2 | XP_005260644.1 | 8 - 9 | 1397 | 124 | ||
| XM_005260590.2 | XP_005260647.1 | 8 - 9 | 1397 | 124 | ||
| XM_011529090.2 | XP_011527392.1 | 8 - 9 | 1325 | 124 | ||
| XM_017028110.1 | XP_016883599.1 | 7 - 8 | 1262 | 124 | ||
| GenBank mRNA | AF041260.1 | - | 7 - 8 | 1261 | 124 | |
| AK294135.1 | - | 3 - 4 | 381 | 124 | ||
| AK295318.1 | - | 5 - 6 | 796 | 124 | ||
| AK315220.1 | - | 7 - 8 | 1482 | 124 | ||
| BC126346.1 | - | 7 - 8 | 1193 | 8992062 | 124 | |
| BC126348.1 | - | 7 - 8 | 1193 | 8992064 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000960.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294135.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013185.hg.1 | ENST00000371435.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013185.hg.1 | ENST00000395961.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013185.hg.1 | ENST00000448710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013185.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013185.hg.1 | TC20000960.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | TC2000009516.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166495.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371435.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000371440.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000395961.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000422805.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | ENST00000448484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009516.hg.1 | NM_003657.3 | Refseq |

BCAS1
Genomic Map