| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
ENST00000322229.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
ENST00000357345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
ENST00000382491.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
ENST00000382499.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
ENST00000433931.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
NM_001160302.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
NM_001160306.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
NM_003895.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
NM_203446.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21004508.hg.1 |
TC21000400.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000357345.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000382491.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000382499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000429236.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000433931.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000630077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_001160302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_001160306.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_003895.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_203446.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
TC2100007964.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000357345.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000382491.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000382499.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000429236.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000433931.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
ENST00000630077.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_001160302.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_001160306.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_003895.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100153657.hg.1 |
NM_203446.2 |
Refseq |