Assay Details
| Gene Symbol: CYB5R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016230.3 | NP_057314.2 | 3 - 4 | 461 | 82 | |
| GenBank mRNA | AF169803.1 | - | 2 - 3 | 233 | 82 | |
| AK313146.1 | - | 3 - 4 | 434 | 82 | ||
| BC025380.1 | - | 3 - 4 | 428 | 4829042 | 82 | |
| DA274558.1 | - | 3 - 4 | 466 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000774.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of oxygen
generation of precursor metabolites and energy
NADP metabolic process
superoxide metabolic process
bicarbonate transport
insulin secretion
glucose homeostasis
response to antibiotic
cell development
oxidation-reduction process
NADPH-hemoprotein reductase activity
cytochrome-b5 reductase activity, acting on NAD(P)H
NAD(P)H oxidase activity
oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
heme binding
metal ion binding
FAD binding
detection of oxygen
generation of precursor metabolites and energy
NADP metabolic process
superoxide metabolic process
bicarbonate transport
insulin secretion
glucose homeostasis
response to antibiotic
cell development
oxidation-reduction process
NADPH-hemoprotein reductase activity
cytochrome-b5 reductase activity, acting on NAD(P)H
NAD(P)H oxidase activity
oxidoreductase activity, acting on NAD(P)H, heme protein as acceptor
heme binding
metal ion binding
FAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06009273.hg.1 | TC06000774.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009273.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009273.hg.1 | ENST00000369681.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009273.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009274.hg.1 | TC06000774.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009274.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009274.hg.1 | ENST00000369681.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06009274.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199056.hg.1 | TC0600014148.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | ENST00000479164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014148.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | ENST00000369679.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | ENST00000369681.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | NM_016230.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199057.hg.1 | TC0600014148.hg.1 | Clariom D TC |

CYB5R4
Genomic Map