Assay Details
| Gene Symbol: IDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289910.1 | NP_001276839.1 | 4 - 5 | 460 | 72 | |
| NM_001290114.1 | NP_001277043.1 | 2 - 3 | 435 | 72 | ||
| NM_002168.3 | NP_002159.2 | 4 - 5 | 693 | 72 | ||
| GenBank mRNA | AK222916.1 | - | 4 - 5 | 607 | 72 | |
| AK294148.1 | - | 4 - 5 | 460 | 72 | ||
| AK299987.1 | - | 2 - 3 | 349 | 72 | ||
| AK312627.1 | - | 4 - 5 | 657 | 72 | ||
| AK316388.1 | - | 4 - 5 | 534 | 72 | ||
| BC009244.2 | - | 4 - 5 | 595 | 2959540 | 72 | |
| BC071828.1 | - | 4 - 5 | 594 | 6090792 | 72 | |
| X69433.1 | - | 4 - 5 | 615 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001841.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | ENST00000330062.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | ENST00000539790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | ENST00000559482.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15017896.hg.1 | TC15001841.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500178578.hg.1 | TC1500010422.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000330062.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000540499.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000559482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | ENST00000560061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001289910.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_001290114.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010422.hg.1 | NM_002168.3 | Refseq |

IDH2
Genomic Map