Assay Details
| Gene Symbol: SLC15A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005073.3 | NP_005064.1 | 7 - 8 | 629 | 100 | |
| GenBank mRNA | AF043233.1 | - | 7 - 8 | 608 | 100 | |
| AY634368.1 | - | 7 - 8 | 608 | 100 | ||
| BC096327.1 | - | 7 - 8 | 583 | 40008278 | 100 | |
| BC096328.3 | - | 7 - 8 | 583 | 40008279 | 100 | |
| BC096329.3 | - | 7 - 8 | 583 | 40008280 | 100 | |
| EU544224.1 | - | 6 - 7 | 552 | 100 | ||
| U13173.1 | - | 7 - 8 | 608 | 100 | ||
| U21936.1 | - | 7 - 8 | 608 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000824.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009193.hg.1 | ENST00000376503.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009193.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13009193.hg.1 | TC13000824.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300164049.hg.1 | TC1300009580.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000376503.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000613008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | ENST00000618992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009580.hg.1 | NM_005073.3 | Refseq |

SLC15A1
Genomic Map